Ion Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78146-1 78146-2A 78146-3 78146-5A 78146-7 78146-O 78146-OO 78146-SS
102098-836EA 62.03 CAD
102098-836 102098-838 102098-840 102098-846 102098-848 102098-850 102098-852 102098-854
Ion Tweezers, Electron Microscopy Sciences
Forceps
These tweezers feature a special surface coating which releases silver ions. These ions act as an anti-bacterial agent which helps the user from contamination of any pathogens.
Unlike other methods, the use of ion technology does not give rise to any resistance. Consequently, it represents a risk-free method of guaranteeing hygiene, even at workstations where instruments are used by several different people. Made from finest stainless steel grade, these tweezers are perfect companions for your daily use in the laboratory. (We recommend that safety gloves still be used whenever handling any infectious materials).
Style SS measures 135 mm (5 1/3”) Long. M = 8 mm (1/3”) N = 1.5 mm (1/16”)    P = 82 mm (3 ¼”). Style 1 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P =  50 mm (2”). Style 3 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 50 mm (2”). Style 3C measures 110 mm (4 1/3”) Long. M = 9 mm (1/3”) N = 1.5 mm (1/16”)   P = 43 mm (1 2/3”). Style O measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 51 mm (2”).  Style OO measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 2.5 mm (1/10”) P = 46 mm (1 ¾”) U = 0.5 mm (½0”).  Style 2A measures 120 mm ( ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 35 mm (1 3/8”) U = 2 mm (1/12”).  Style 5 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 30 mm (1 1/16”). Style 5A measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).  Style 7 measures 120 mm (4 ¾”) Long. M = 11 mm (½”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).