EMS Wafer Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78411-3WF 78412-39S2 78414-4WF 78426-96 78414-43WFG 78420-8WF 78422-8WNY 78411-38FG 78412-39S3 78414-4WFC 78414-44WF 78414-48WF 78410-2W 2WFG.SA 3WL.SA 4WL.SA 5WF.SA 5WL.SA
100494-860EA 75.43 CAD
100494-860 100494-862 100494-874 100494-882 100494-868 100494-876 100494-878 100494-858 100494-864 101414-018 100494-870 100494-872 100494-850 76499-512 76499-520 76499-532 76499-542 76499-544
EMS Wafer Tweezers, Electron Microscopy Sciences
Forceps
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel.
Order Now
SPECIFICATIONS
Environmentally Preferable