EMS Zirconia Ceramic Tip Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78127-2A 78127-74 78127-71 78127-72 78127-7 78127-73 78127-2A-T 78127-71-T 78127-72-T 78127-73-T 78127-74-T 78127-7-T 72ZJ.SA
100494-604EA 157.42 CAD
100494-604 100494-614 100494-608 100494-610 100494-606 100494-612 76376-114 76376-116 76376-118 76376-120 76376-122 76376-124 76499-592
EMS Zirconia Ceramic Tip Tweezers, Electron Microscopy Sciences
Forceps
These tweezers have Zirconia ceramic precision tips.
  • Light weight
  • Aluminum handles
Surface resistively 10E12 ohm, Contamination free, high temperature (1000 °C). All styles have length of 134 mm (5¹/₄").