Plastic Tweezers and Tweezers Accessories, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78310-05 78311-5A2 78310-51 78310-71 78311-4A 78310-07 78310-06
100494-684EA 32.6 CAD
100494-684 100494-702 100494-692 100494-696 100494-700 100494-688 100494-686
Plastic Tweezers and Tweezers Accessories, Electron Microscopy Sciences
Forceps
Plastic wafer tweezers are low cost and designed for ease of use. Available in glass filled Delrin and acid resistant fluorocarbon or polyethylene.