EMS Fiber Tip Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78085-CF 78010-CP 78014-CP 78095-CP 78014-RFZ 78094-CFZ 78094-CF 78095-CF 78080-CF 78080-CP 78094-CP 78092-CP 78012-RF 78010-RFZ 78012-CP 78084-CP 78085-CP 78090-CFZ 78090-CP 78010-RF 78014-RF 78092-CF 78016-RF 78090-CF 78080-CFZ
100502-878EA 80.26 CAD
100502-878 101413-986 101413-990 101414-006 100499-838 100499-844 100502-280 100502-282 100502-874 101413-994 101414-004 101414-002 100494-594 100499-834 101413-988 101413-996 101413-998 100499-840 101414-000 100494-592 100494-596 100494-602 100494-598 100494-600 102098-814
EMS Fiber Tip Tweezers, Electron Microscopy Sciences
Forceps
Tweezers with anti-magnetic steel handles and synthetic fiber tips.
Synthetic Fiber has high-performance Plastic type CP - PEEK/CF30 (Polyetheretherketone reinforced with 30 wt% carbon fiber). Very hard, high tensile and flexural strength. High heat- 260 – 300°C – resistance to thermal aging. Excellent chemical resistance. ESD safe – low surface resistivity (102 Ohm).
Engineering Plastic type CF - PA66/CF30 (Polyamide 66 reinforced with 30 wt% carbon fiber) is heat stable(-40°C to 190°C). Rigidity, excellent tensile strength and flexural strength. Good chemical resistance (oils, greases,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable