SP Bel-Art No-Wire Submersible Plastic Test Tube Grip Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18749-0001 F18749-0000 F18749-0002
20907-938CS 536.12 CAD
20907-938 20907-940 20907-942 490012-354 490012-476
SP Bel-Art No-Wire Submersible Plastic Test Tube Grip Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Retainer fingers hold tubes firmly in the rack plates with no fear of dislodging, making them perfect for both decanting and water bath work.
  • Fingers allow simultaneous decanting to save valuable time
  • Specially formulized plastic allows the rack to submerge in water baths
  • Fingers secure tubes while in water baths to eliminate the risk of diluting or spilling
  • Features end plates for labeling as well as numbered and lettered rows for tube identification
  • Fold up sides lock firmly into the grip decks; assembly takes only seconds
  • Unloaded racks are stackable
Tube size indicated is for grip function, smaller tubes may fit for storage purposes.