ACTERO™ MediaBox, Foodchek
Supplier: Avantor
CA89522-688EA 338.8 CAD
CA89522-688 CA89522-678 CA89522-680 CA89522-682 CA89522-684 CA89522-686 CA89522-696 CA89522-698 CA89522-700 CA89522-702 CA89522-704 CA89522-706 CA89522-708 CA89522-710 CA89522-712 CA10127-232 CA10127-234 CA10146-834 CA10146-836 CA10146-838 CA10146-840 CA10146-842 CA10146-844 CA10146-846 CA10146-848 CA10146-850 CA10159-100 CA10159-102 CA10159-104 CA10159-106
ACTERO™ MediaBox, Foodchek
Media for Microbiology Prepared Media for Microbiology
The Actero™ MediaBox™ is the solution for laboratories that require liquid culture media on-demand
While Actero Universal Enrichment media represent fast-acting and established formulations that can be used for standard testing protocols employed in today’s labs, the Patent Pending Actero Elite Enrichment Media represent enhanced formulations that further reduce the required enrichment time to an industry-leading low.
Get even faster time-to-results and accuracy when you combine Actero Enrichment Media with the FoodChek™ MICT Reader and Assay
Delivering fresh, sterile media in an easy-to-handle, stackable, storage box, Actero™ MediaBox™ provides affordable, ready-to-use liquid culture media in 5 L and 10 L sizes, with a 20 L option available upon request.