Alpine Fire Assay Cupels, MgO, Anachemia
Supplier: Anachemia Mining
029-730-03 029-730-04 029-730-06 029-730-07 029-730-09 029-730-11 029-730-12
CA11022-132CS 318.75 CAD
CA11022-132 CA11022-134 CA11022-136 CA11022-138 CA11022-140 CA11022-142 CA11022-144 80704-004 CA11022-141
Alpine Fire Assay Cupels, MgO, Anachemia
Crucibles
Alpine cupels are made from high purity magnesium oxide and offer superior mechanical strength free from cracking and pitting
  • High-purity material
  • Superior mechanical strength
  • Consistent low losses
These cupels offer consistent low losses of precious metals and are continually tested to ensure product quality remains at the highest level.