Tri-Pour Plastic Beakers, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
PB5935-250 PB5935-400 PB5935-800
89011-784CS 85.21 CAD
89011-784 89011-786 89011-788
Tri-Pour Plastic Beakers, Medegen Medical Products
Beakers
These Tri-pour beakers have a pour spout on three sides with easy to read graduations on the outside.
  • Made from high-strength polypropylene
  • Non sterile
Easy to use to transfer liquids.