E.Z.N.A. FFPE RNA Kit, Omega Bio-tek

Supplier: OMEGA BIO-TEK

R6954-00 R6954-01 R6954-02
CA89167-336EA 105.65 CAD
CA89167-336 CA89167-338 CA89167-340
E.Z.N.A. FFPE RNA Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The E.Z.N.A.® FFPE RNA Kit is specially designed for isolation of total RNA from formalin-fixed, paraffin-embedded tissue sections


This method can also be used for the preparation of total RNA from fibrous samples such as heart and muscles tissue. The sample is first processed in special buffer by heating to deactivate RNase’s and deparaffinization. Then the lysate is treated with Proteinase K to digest proteins. The cleared lysate is then applied to a HiBind RNA column to capture RNA. After two quick wash steps, purified RNA is eluted with water. While E.Z.N.A.® FFPE RNA Kit is optimized to minimize the effect of the formaldehyde modification, it is not recommended to use the RNA purified from this kit for downstream applications that requires full length RNA.

Order Now

SPECIFICATIONS

Description E.Z.N.A. FFPE RNA Kit
Environmentally Preferable
Application
Target molecule RNA
Refine target molecule Total RNA
Sample type FFPE tissues
Method / Format Column (silica-based)
Prep size Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR