SP Bel-Art® Churn Sample Splitters, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37805-0004 F37805-0008 F37805-0014
47727-668EA 1556.95 CAD
47727-668 47727-670 47727-674
SP Bel-Art® Churn Sample Splitters, Bel-Art Products, a part of SP
Samplers Solid Samplers
Provides an efficient method of preparing subsamples of collected water-sediment mixtures from stream flows for water quality analysis.
  • Polyethylene construction
  • Polypropylene spring-loaded spigot
  • Convenient carrying handle
  • Replaceable lid and spigot
Water sediment samples are poured into the sample splitter where they are composited into one representative cross section sample of the stream flow
The polyethylene sample splitter consists of a thick walled cylindrical vessel with a disc churn. A center handle extends through the cover and a locating notch keeps it oriented in one direction. A small polypropylene spring-loaded spigot drains the subsamples into bottles for delivery to the water quality laboratory.
A carrying handle makes carrying the full or empty vessel easy. Lid and spigot are replaceable.
This composite can now be split into the required four to sixteen representative subsamples.