MicroFunnel™ Filter Funnels with Polycarbonate Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
FMFNL1050 FMFNL3020
CA97058-848PK 498 CAD
CA97058-848 CA97058-850
MicroFunnel™ Filter Funnels with Polycarbonate Membrane, Cytiva (Formerly Pall Lab)
Funnels Filtration Funnels Micro Filter Funnels
The MicroFunnel™ Filter Funnels are ideal for concentrating organisms in fluid samples.
  • Polycarbonate membrane
  • Smooth surface
  • Easy elution
  • Disposable
The flat, smooth surface allows for easy elution of organisms or particulate and the ready to use, disposable design eliminate sample cross contamination. The collection of organisms or particulate on one planar surface is ideal prior to electron microscopy (SEM) or light microscope (LM) techniques. Funnels feature a polycarbonate membrane. The unique squeeze separation makes membrane retrieval easy.