SP Bel-Art Stainless Steel Micro Spatulas, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36700-0000 H36703-0000 H36704-0000
47750-084PK 76.48 CAD
47750-084 47750-086 47750-088
SP Bel-Art Stainless Steel Micro Spatulas, Bel-Art Products, a part of SP
Spatulas
These micro spatulas are made of stainless steel rod with flat ends coated in brown Teflon® FEP, which complies with FDA regulations governing direct food contact applications.
  • FEP-coated stainless steel construction
  • Coating provides anti-stick, corrosion-resistant surface
Available with straight or tapered blade, the ends measure 7.9W×50L mm (⁵⁄₁₆×2").