PYREX® Heavy Duty Conical Centrifuge Tubes, Graduated, Corning
Supplier: Corning
8140-12 8140-40
89091-146CS 744.84 CAD
89091-146 89091-378
PYREX® Heavy Duty Conical Centrifuge Tubes, Graduated, Corning
Tubes Centrifuge Tubes
PYREX® borosilicate glass, conical, heavy duty, beaded rim.
  • Tubes 12 ml are graduated upward from tip in 0.1 ml divisions
  • Tubes 40 ml are graduated in 0.5 ml divisions to 10 ml and from 10 to 40 ml in 1 ml divisions
The tubes are of heavy wall construction, for greater mechanical strength, with durable, white enamel graduations.
In order to conform to outside diameter and overall length limitations imposed by centrifuge rotors and supports, it has been necessary to reduce the capacity of these heavy duty tubes.