PYREX® Capillary Melting Point Tubes, Corning
Supplier: Corning
9530-1 9530-3 9530-4 9530-2
32311-080CS 647.83 CAD
32311-080 32311-084 32311-086 89091-084
PYREX® Capillary Melting Point Tubes, Corning
Tubes Melting Point Tubes and Capillaries
Quality glassware at an affordable price.
  • Disposable
  • Available open-ended on both sides or closed on one side
  • Made from no. 7740 glass
All tubes have a 0.2 to 0.25 mm wall range.