CDC861T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
E61M016 E61M016
CA89501-080EA 3273.49 CAD
CA89501-080 89501-080
CDC861T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Electrodes Conductivity Sensors
The Radiometer CDC861T is a 4-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and glass body
The CDC861T sensor's 4 platinum ring design and glass body ensures results in general aqueous and non-aqueous media, including very strong acids and bases or saline media, over several decades of conductivity with a single cell, using just one calibration. It has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters.