SP Bel-Art Graduated Griffin Low-Form Beakers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F26213-0000 F26211-0000 F26215-0000 F26210-0000 F26219-0000 F26212-0000 F26217-0000 F26214-0000 F26216-0000
30617-156CS 429.34 CAD
30617-156 47750-436 13197-468 53502-276 30617-154 47750-450 21812-906 47750-442 47750-434
SP Bel-Art Graduated Griffin Low-Form Beakers, Bel-Art Products, a part of SP
Beakers
Griffin low-form polypropylene plastic beakers are translucent, chemical resistant, and will stack without sticking together.
  • Polypropylene construction
  • Low-form design
  • Translucent
  • Chemical resistant
  • Stackable
  • Autoclavable
Raised numeric graduations show approximate volume (± 5%).
All sizes are steam autoclavable at 121ºC (250ºF) and microwavable.