IntelliCAL™ ISECL181 Chloride Ion Selective Electrode (ISE), Hach
Supplier: Hach
IntelliCAL™
ISECL18103 ISECL18103
89501-100EA 2526.07 CAD
89501-100 CA89501-100
IntelliCAL™ ISECL181 Chloride Ion Selective Electrode (ISE), Hach
Electrodes ISE Electrodes
IntelliCAL™ ISECL181 is a digital, combination chloride ion selective electrode (ISE) with a non-refillable, double junction reference and built-in temperature sensor.
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include dry storage and fast response time. No replacement membranes.
The IntelliCAL ISECL181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISECL181 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications. The ISECL181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Temperature range:
Continuous use; 5 to 50 °C
Ordering information: Includes IntelliCAL ISECL181 combination Chloride ISE with integrated temperature sensor, calibration certificate, and Basic User Manual.