gYEAST Mini Genomic DNA Reagent Kits, IBI Scientific
Supplier: IBI Scientific
The gYEAST Genomic DNA Kit offers a simple and gentle reagent DNA precipitation method for isolating high molecular weight genomic, mitochondrial or viral DNA from Saccharomyces cerevisiae and a variety of other yeast and fungus species
This highly versatile, solution-based system offers a convenient procedure with minimal hands on time. The provided Sorbitol Buffer, when combined with β-mercaptoethanol, zymolase, or lyticase, will efficiently lyse yeast and other fungus species cell walls consisting of chitin and polysaccharides. The extracted DNA is suitable for use in PCR, AFLP, RFLP/PADP, Southern Blotting, and Real-Time PCR.
Sample size: up to 2x108 of yeast or other fungus species
Ordering information: Each kit includes Sorbitol Buffer, Cell Lysis Buffer, Protein Removal Buffer, and DNA Hydration Buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0).