E.Z.N.A.® Water DNA Kit, Omega Bio-tek
Supplier: OMEGA BIO-TEK CA
D5525-00 D5525-01
CA95043-150EA 42.23 CAD
CA95043-150 CA95043-152
E.Z.N.A.® Water DNA Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Water DNA Kit is formulated to isolate high-purity cellular DNA from water samples typically containing humic acid and inhibitors of PCR.
  • Reliability – Reproducible DNA purification from variety of sample sources
  • Quality – Inhibitor Removal Technology provides efficient PCR inhibitor removal
  • High Yield – Efficient purification of DNA from even specialized samples
This kit uses a novel and proprietary method to isolate genomic DNA from variety of environmental samples. This kit has been successfully used to isolate DNA from tough-to-lyse bacteria, fungi, and algae that inhabit a range of samples including Tap water, Lake, River and Sewage samples. Isolated DNA can be used for most downstream applications including PCR, Southern blot, SNP analysis.