Boekel Scientific Tight Temperature Incubators
Supplier: Boekel
135000 135000-2
10027-266EA 3729.12 CAD
10027-266 10027-268
Boekel Scientific Tight Temperature Incubators
Incubators Standard Incubators
The Boekel TTT Incubator is specifically designed for maximum temperature stability and uniformity while using forced air convection for those temperature sensitive applications. Ideal for water/wastewate protocols or any microbiology and biology protocol requiring tight uniformity and temperature.
  • Large viewing window
  • Easy to use digital control system
  • User configurable alarm and timer
  • Stackable
  • Removable shelf
  • Designed for benchtops with limited space
Using mechanical convection and an optimized configuration it is ideal for any small sample temperature sensitive process.
Delivery information: One removeable shelf is included with each unit.