E-Z 96 DNA Plates, Omega Bio-tek
Supplier: OMEGA BIO-TEK CA
BD96-02 BD96-01
CA95030-224CS 4685.63 CAD
CA95030-224 CA95030-226
E-Z 96 DNA Plates, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
E-Z 96 plates are silica glass fiber plates that can bind up to 50 μg of genomic DNA or 20 μg of plasmid DNA per prep
Each well has a capacity of 1 mL and the plates are semi skirted. E-Z 96 DNA plates cab be process in vacuum manifolds and/or by centrifugation. Omega Bio-tek Columns are compatible with following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, Dneasy Plant DNA.