IntelliCAL™ ISEF121 Fluoride Ion Selective Electrode (ISE), Hach
Supplier: Hach
ISEF12103 ISEF12103
89501-104EA 3521.36 CAD
89501-104 CA89501-104
IntelliCAL™ ISEF121 Fluoride Ion Selective Electrode (ISE), Hach
Electrodes ISE Electrodes
IntelliCAL™ ISEF121 is a digital, combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include dry storage and fast response time. No replacement membranes.
The IntelliCAL ISEF121 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISEF121 is ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications. The ISEF121’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Temperature range:
Continuous use: 5 to 50 °C
Ordering information: Includes IntelliCAL ISEF121 combination Fluoride ISE with integrated temperature sensor, calibration certificate, and Basic User Manual.