CDC566T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
E61M010 E61M010
89501-074EA 2801.28 CAD
89501-074 CA89501-074
CDC566T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Electrodes Conductivity Sensors
The Radiometer CDC566T Conductivity Cell is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), robust epoxy body and integrated temperature sensor
  • The right conductivity cell for your application
It is important to choose a conductivity cell with the right construction and geometry for your particular application and working conditions. The Radiometer Analytical line offers a variety of conductivity cells to meet a large number of applications.
The CDC566T sensor's 4-pole platinum ring design ensures results in general aqueous and non-aqueous media over several decades of conductivity with a single cell, using just one calibration. The epoxy body of the probe can be easily removed for rinsing, which makes the cell ideal for measurements in harsh conditions. The CDC566T has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M021 for the CDC536T (3-pole), compatible for use with Hach sensION+ benchtop conductivity meters.
Ordering information: Includes Radiometer Analytical CDC566T conductivity cell with Certificate of Conformity.