gBAC Mini Genomic DNA Kits, IBI Scientific
Supplier: IBI Scientific
IBI gBAC Mini DNA Bacteria Kit is optimized for genomic and viral DNA purification from Gram (-) negative and Gram (+) positive bacterial cells
- Sample Size: up to 1×109gram (+) or gram (-) bacterial cells
- Expectant Yield: up to 30µg DNA
- Operation Time: 30min or less
- Elution Volume: 50-200µl
- Free sample available upon request
The gBAC Mini Genomic DNA Kit has been optimized for purifying total DNA (including genomic and viral DNA) from gram negative and gram positive bacterial cells. The provided gram positive buffer, when combined with lysozyme, will efficiently lyse bacterial cell walls consisting of peptidoglycan. Chaotropic salt is then used to further lyse cells and degrade protein, allowing DNA to bind to the glass fiber matrix of the spin column. Contaminants are removed using a wash buffer (containing ethanol) and the purified genomic DNA is eluted using a low salt elution buffer or TE. The entire procedure can be completed in one hour without phenol│chloroform extraction or alcohol precipitation. The purified DNA is suitable for PCR and other enzymatic reactions.
The quality of the gBAC Mini DNA Bacteria Kit is tested on a lot-to-lot basis by isolating DNA from E. coli (1x109) culture (OD600=1.3, 1 ml) harvested by centrifugation at 16,000 xg for one minute. 10 µl from a 50 µl eluate of purified DNA is analyzed by electrophoresis on a 1% agarose gel.
Specifications
Format: Spin Column
Binding Capacity: 50 µg
Sample Type: Gram (+) positive and Gram (-) negative bacterial cells,blood and biological fluids such as plasma, urine etc.
Sample Concentration: 1x109bacterial cells 200 µl blood 1 ml urine, plasma
Enzyme: Lysozyme Proteinase K
Typical Yield: 37 µg from 1 x 109 E. coli 10-15 µg from 1 x 109 Bacillus subtilis
Elution Volume: 30-200 µl
Operation Time: <30 minutes
Ordering information: Each kit comes with Gram (+) Buffer, GT Buffer, GB Buffer, W1 Buffer, Wash Buffer, Elution Buffer, GD Columns, and 2 mL Collection Tubes.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...