SP Bel-Art Stainless Steel Spoons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36729-0021 H36729-0030 H36729-0015 H36729-0018 H36729-0025 F36730-0015 F36730-0030
13197-382PK 157.92 CAD
13197-382 13197-386 13197-378 13197-380 13197-384 47727-684 47727-686 47727-690
SP Bel-Art Stainless Steel Spoons, Bel-Art Products, a part of SP
Spoons
These double-ended, 304 stainless steel sampling spoons have a solid, rounded shaft.
  • Double-ended design
  • Stainless steel construction
  • Optional FEP coating provides non-stick surface
Available in plain polished stainless steel or coated with Fluo-Kem® Teflon® FEP, which complies with FDA regulations governing direct food contact applications.