ClipTip® Pipette Tips, 12.5, Thermo Scientific
Supplier: Thermo Scientific
94410040 94410043 94420043 94410060 94410063 94420063
10030-778CS 787 CAD
10030-778 10030-780 10030-782 76289-892 76289-894 76289-900
ClipTip® Pipette Tips, 12.5, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® 12.5 pipette tips feature a unique ‘Snap and Lip’ seal technology to attach and seal the tips on each channel with minimal tip attachment and ejection force.
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Available in nonsterile, sterile, and filtered formats
  • Each tip insert has ink-jetted lot and volume information
Convenient color keys make it easy to match the color of the E1-ClipTip® pipettor with the corresponding ClipTip® pipette tip rack insert.
ClipTip® 12.5 pipette tips are suitable forMore Product Information