The VWR® 250 Homogenizer unit is a perfect fit between the VWR® 200 Homogenizer unit and the benchtop VWR® Homogenizer units.
- Versatile handheld homogenizer unit
- Ideal for processing volumes from 0.03 ml to 5L
- High-torque motor with variable speed adjustment of 10,000 to 30,000 rpm
- Use handheld or post mount to a stand
- VWR® Generator probe sold separately
This unit is lightweight enough to be handheld for convenience, yet offers extra torque for processing difficult samples
With VWR® generator probes ranging in diameter from 5mm to 30mm, it can process samples from 0.03 mL to 5L (higher sample volumes dependent on viscosity).
A separate ON/OFF switch effortlessly allows for the dial knob speed control to remain at an ideal homogenizing speed. Combining the power of a ¾hp, 576 watt motor with smooth variable speed control, the unit is capable of easily handling a wide range of sample sizes.
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. A VWR® Generator probe(s) are required for use with the VWR® 250 Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes). Optional accessory items include; VWR® 250 Homogenizer Stand Assembly (with 10" open U-shaped base, 24" post, motor cross rod, and post clamp), VWR® 250 Homogenizer Small Footprint Stand Assembly (with 6" padded base, 24" post, motor cross rod, and post clamp). All accessories are sold separately.