Quartz SUPRASIL Macro/Semi-Micro Cell, PerkinElmer
Supplier: Perkin Elmer
B0631132 B0631132
CA97013-452EA 1175.08 CAD
CA97013-452 97013-452
Quartz SUPRASIL Macro/Semi-Micro Cell, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
The Quartz SUPRASIL Macro/Semi-Micro Cell has a 4 x 4 mm light path with 4 windows.
  • 4 windows for excellent visibility
  • Interior width: 4 mm
  • Cell volume: 500 μL
Standard rectangular macro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
Ordering information: Cell requires stirrer fleas.