VWR® MaX Homogenizing Packages
Supplier: VWR International
10032-484 10032-764 10032-766
10032-484KT 5023.42 CAD
10032-484 10032-764 10032-766
VWR® MaX Homogenizing Packages
Homogenizers Handheld Homogenizers
The VWR® Standard MaX-Homogenizing Package is ideal for homogenizing in various tube sizes, from 10 mL to 50 mL.
  • Stand assembly included
  • Fast and efficient sample breakdown
The VWR® Premium MaX-Homogenizing Package is supplied complete with stand assembly for standard hands-free homogenizing in 10 mL to 50 mL tubes, and medium to large beakers, all without having to hold the homogenizer unit
The VWR® Deluxe MaX-Homogenizing Package is perfect for homogenizing in 10 mL to 50 mL tubes, and medium to large beakers. Individual samples as large as ½” in diameter can be processed. Various types of samples can be homogenized fast and complete, even tough or stringy sample types. Designed for use with sample volumes of 1.5 mL to 2.5L. Both 10mm and 20mm sawtooth generator probes provide fast and efficient sample breakdown.
The package enables a broad processing capability of all types of samples, even tough or stringy. A typical volume range of 1.5 mL to 30 mL can be efficiently homogenized. A 10mm sawtooth generator probe provides high shearing for compete sample breakdown.
Individual samples as large as ½” in diameter can be processed. Various types of samples can be homogenized fast and complete, even tough or stringy sample types. Designed for use with sample volumes of 1.5 mL to 2.5L. Both 10mm and 20mm sawtooth generator probes provide fast and efficient sample breakdown.
Certifications: CE marked.
Ordering information: The VWR® Standard MaX-Homogenizing Package (10032-484) includes VWR® 200 Homogenizer unit and 10x115mm saw-tooth generator probe. The VWR® Deluxe MaX-Homogenizing Package (10032-764) includes VWR® 250 Homogenizer unit, 10x115mm saw-tooth generator probe, and 20x200mm saw-tooth generator probe. The VWR® Premium MaX-Homogenizing Package (10032-766) includes VWR® 250 Homogenizer unit, 10x115mm saw-tooth generator probe, 20x200mm saw-tooth generator probe, and a stand assembly with a 15.2x15.2 cm (6x6") footprint.