Diindium trisulfate, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Diindium tris(sulphate), Indium(III) sulphate, Diindium tris(sulfate), Indium (III) sulfate, Indium (III) sulphate, Indium(III) sulfate

M-Clarity™ quality level = MQ200

8.43982.0025 8.43982.0100 8.43982.0025 8.43982.0100
EM8.43982.0025CS 18013.58 CAD
EM8.43982.0025 EM8.43982.0100 CA8.43982.0025 CA8.43982.0100
Diindium trisulfate, anhydrous for synthesis, Sigma-Aldrich®
Diindium trisulphate
Formula: O₁₂In₂S₃
MW: 517.83 g/mol
Melting Pt: 585
Density: 3.438 g/cm³ (20 °C)
MDL Number: MFCD00011059
CAS Number: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978

Order Now

Specification Test Results

Molar Mass 517.83 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR