Propanephosphonic acid cyclic anhydride 50% in 2-MeTHF (2-methyltetrahydrofuran), T3P® for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide

M-Clarity™ quality level = MQ200

8.43943.0100 8.43943.0025 8.43943.0050 8.43943.0100
EM8.43943.0025CS 22161.08 CAD
EM8.43943.0025 EM8.43943.0050 EM8.43943.0100 CA8.43943.0025 CA8.43943.0050 CA8.43943.0100
Propanephosphonic acid cyclic anhydride 50% in 2-MeTHF (2-methyltetrahydrofuran), T3P® for synthesis, Sigma-Aldrich®
Propanephosphonic acid cyclic anhydride
Formula: C₉H₂₁O₆P₃
MW: 318.19
MDL Number: MFCD00006583
CAS Number: 68957-94-8
EINECS: 422-210-5
UN: 2924
ADR: 3 (8),II

Order Now

Specification Test Results

Boiling Point 78 - 80 °C (solvent)
Flash Point -11 °C (solvent)
Density 1.039 g/cm3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR