Polyethylene glycol diacid, M.W. 600 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Poly(ethylene glycol) bis(carboxymethyl) ether

M-Clarity™ quality level = MQ200

8.43912.0250 8.43912.1000 8.43912.0250 8.43912.1000
EM8.43912.0250CS 4542.5 CAD
EM8.43912.0250 EM8.43912.1000 CA8.43912.0250 CA8.43912.1000
Polyethylene glycol diacid, M.W. 600 for synthesis, Sigma-Aldrich®
Polyethylene glycol diacid
Formula: (C₂H₄O)nC₄H₆O₅
MDL Number: MFCD00132002
CAS Number: 39927-08-7

Order Now

Specification Test Results

Colour (visual) colourless
Appearance of substance (visual) liquid
Acid value 160 - 195
Water (K. F.) ≤ 5.0 %
Refractive index (n 20°/D) 1.460 - 1.480


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR