[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
[2-(2-Methoxyethoxy)ethoxy]acetic acid, 3,6,9-Trioxadecanoic acid, 2-(2-(2-Methoxyethoxy)ethoxy)acetic acid
M-Clarity™ quality level = MQ200
8.43910.0050
8.43910.0250
8.43910.0050
8.43910.0250
EM8.43910.0050CS
13502.68
CAD
EM8.43910.0050
EM8.43910.0250
CA8.43910.0050
CA8.43910.0250
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) for synthesis, Sigma-Aldrich®
[2-(2-Methoxyethoxy)ethoxy]acetic acid (3,6,9-Trioxadecanoic acid)
Formula:
C₇H₁₄O₅ MW: 178.1831 g/mol |
MDL Number:
MFCD00044098 CAS Number: 16024-58-1 EINECS: 240-162-1 UN: 3265 ADR: 8,III |
Specification Test Results
Molar Mass | 178.18 g/mol |
Boiling Point | 140 °C (2 mbar) |
Flash Point |
|
Density | 1.157 g/cm3 |
pH | 1.8 (100 g/l, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...