5-(4-Fluorophenyl)-3-pyridinecarboxylic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43818.0005 8.43818.0025 8.43818.0005 8.43818.0025
EM8.43818.0005CS 73882.38 CAD
EM8.43818.0005 EM8.43818.0025 CA8.43818.0005 CA8.43818.0025
5-(4-Fluorophenyl)-3-pyridinecarboxylic acid for synthesis, Sigma-Aldrich®
5-(4-Fluorophenyl)-3-pyridinecarboxylic acid
Formula: C₁₂H₈FNO₂
CAS Number: 364064-17-5

Order Now

Specification Test Results

Melting Point 215 °C
Molar Mass 217.20 g/mol
Density 0.5 g/cm3 (20 °C)
pH 5.7 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR