Hydrazine dihydrochloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hydrazine Dihydrochloride, Hydrazinium dichloride
M-Clarity™ quality level = MQ200
8.43816.0100
8.43816.1000
8.43816.0100
8.43816.1000
EM8.43816.0100CS
3128.4
CAD
EM8.43816.0100
EM8.43816.1000
CA8.43816.0100
CA8.43816.1000
Hydrazine dihydrochloride for synthesis, Sigma-Aldrich®
Hydrazine dihydrochloride
Formula:
H₆Cl₂N₂ MW: 104.97 g/mol Melting Pt: 189 Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00064543 CAS Number: 5341-61-7 EINECS: 226-283-2 UN: 3288 ADR: 6.1 III Merck Index: 13,04789 |
Specification Test Results
Solubility | 2700 g/l (20 °C) |
Melting Point | 198 °C (decomposition) |
Molar Mass | 104.97 g/mol |
Bulk Density | 770 kg/m3 |
Density | 1.42 g/cm3 (20 °C) |
pH | 0.5 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...