Sodium propionate zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Propionic acid sodium salt, Sodiumpropionate

MQ200

8.43793.0100 8.43793.0500 8.43793.0100 8.43793.0500
EM8.43793.0100CS 9017.06 CAD
EM8.43793.0100 EM8.43793.0500 CA8.43793.0100 CA8.43793.0500
Sodium propionate zS, Sigma-Aldrich®
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76
Flash Pt: 157
MDL Number: MFCD00002759
CAS Number: 137-40-6
Merck Index: 12,08816

Order Now

Specification Test Results

Solubility 995 g/l (20 °C)
Melting Point 285 - 286 °C
Molar Mass 96.06 g/mol
pH 8 - 9.5 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR