(±)-2-Methylenanthic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.43786.0005 8.43786.0025 8.43786.0005 8.43786.0025
EM8.43786.0005CS 125526.26 CAD
EM8.43786.0005 EM8.43786.0025 CA8.43786.0005 CA8.43786.0025
(±)-2-Methylenanthic acid for synthesis, Sigma-Aldrich®
(±)-2-Methylenanthic acid
Formula: C₈H₁₆O₂
Boiling Pt: 85…87 °C (0.5 mmHg)
Density: 0.9122 g/cm³ (20 °C)
MDL Number: MFCD00142944
CAS Number: 1188-02-9
EINECS: 214-704-2

Order Now

Specification Test Results

Molar Mass 144.22 g/mol
Boiling Point 140 °C (40 hPa)
Flash Point 110 °C
Density 0.92 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR