Linoleic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43483.0025 8.43483.0100 8.43483.8100 8.43483.0025 8.43483.0100 8.43483.8100
EM8.43483.0025CS 32631.74 CAD
EM8.43483.0025 EM8.43483.0100 EM8.43483.8100 CA8.43483.0025 CA8.43483.0100 CA8.43483.8100
Linoleic acid for synthesis, Sigma-Aldrich®
Linoleic acid
Formula: C₁₈H₃₂O₂
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -5 °C
Molar Mass 280.45 g/mol
Boiling Point 230 °C (21 hPa)
Flash Point 110 °C
Density 0.903 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR