DL-Tartaric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: (2R,3R)-rel-2,3-Dihydroxysuccinic acid, Racemic Acid

M-Clarity™ quality level = MQ200

8.43377.0100 8.43377.0500 8.43377.0100 8.43377.0500
EM8.43377.0100CS 7138.44 CAD
EM8.43377.0100 EM8.43377.0500 CA8.43377.0100 CA8.43377.0500
DL-Tartaric acid for synthesis, Sigma-Aldrich®
DL-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
MDL Number: MFCD00151254
CAS Number: 133-37-9

Order Now

Specification Test Results

Solubility 215 g/l (25 °C)
Melting Point 198 - 204 °C
Molar Mass 150.09 g/mol
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 210 °C
pH 1.6 (100 g/l, H2O, 25 °C)
Ignition Point 425 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR