Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.43262.0025 8.43262.0100 8.43262.0025 8.43262.0100
EM8.43262.0025CS 12221.3 CAD
EM8.43262.0025 EM8.43262.0100 CA8.43262.0025 CA8.43262.0100
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride for synthesis, Sigma-Aldrich®
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Formula: C₁₂H₈O₆
MDL Number: MFCD00082228
CAS Number: 1719-83-1

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 300 °C
Molar Mass 248.19 g/mol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR