4,4'-Methylenebis(2,6-di-tert-butylphenol) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane

M-Clarity™ quality level = MQ200

8.42857.0010 8.42857.0100 8.42857.0010 8.42857.0100
EM8.42857.0010CS 57927.54 CAD
EM8.42857.0010 EM8.42857.0100 CA8.42857.0010 CA8.42857.0100
4,4'-Methylenebis(2,6-di-tert-butylphenol) for synthesis, Sigma-Aldrich®
4,4'-Methylenebis(2,6-di-tert-butylphenol)
Formula: C₂₉H₄₄O₂
MW: 424.67 g/mol
MDL Number: MFCD00008822
CAS Number: 118-82-1

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (20 °C)
Melting Point 153 - 156 °C
Molar Mass 424.67 g/mol
Boiling Point 217 °C (1 hPa)
Vapor Pressure  - 0.1 hPa (25 °C)
Flash Point 222 °C
Density 0.99 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR