2-Nitrobenzenesulfonyl chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.42593.0025
8.42593.0100
8.42593.0025
8.42593.0100
EM8.42593.0025CS
8524.1
CAD
EM8.42593.0025
EM8.42593.0100
CA8.42593.0025
CA8.42593.0100
2-Nitrobenzenesulfonyl chloride for synthesis, Sigma-Aldrich®
2-Nitrobenzenesulphonyl chloride
Formula:
C₆H₄ClNO₄S Melting Pt: 69.5…69.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007430 CAS Number: 1694-92-4 EINECS: 216-907-1 UN: 3261 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 65 - 67 °C |
Molar Mass | 221.62 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...