1,5-Dichloropentane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.42141.0025
8.42141.0100
8.42141.0025
8.42141.0100
EM8.42141.0025CS
4610.44
CAD
EM8.42141.0025
EM8.42141.0100
CA8.42141.0025
CA8.42141.0100
1,5-Dichloropentane for synthesis, Sigma-Aldrich®
1,5-Dichloropentane
Formula:
ClH₂C(CH₂)₃CH₂Cl Boiling Pt: 63 °C (1013 hPa) Melting Pt: –72.8 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001017 CAS Number: 628-76-2 EINECS: 211-053-6 UN: 1152 ADR: 3,III |
Specification Test Results
Solubility | 0.125 g/l (25 °C) |
Melting Point | -72 °C |
Molar Mass | 141.04 g/mol |
Boiling Point | 180 °C (1013 hPa) |
Vapor Pressure | 1.5 hPa (25 °C) |
Flash Point | 26 °C |
Density | 1.10 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...