2-(4-Hydroxyphenyl)ethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41859.0005 8.41859.0005 8.41859.0025
EM8.41859.0005CS 240919.98 CAD
EM8.41859.0005 CA8.41859.0005 CA8.41859.0025
2-(4-Hydroxyphenyl)ethanol for synthesis, Sigma-Aldrich®
2-(4-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 90…92 °C
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8

Order Now

Specification Test Results

Solubility 124 g/l (25 °C)
Melting Point 91 - 94 °C
Molar Mass 138.17 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR