Tris(2-methylphenyl)phosphine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

M-Clarity™ quality level = MQ200

8.41817.0001 8.41817.0010 8.41817.0001 8.41817.0010
EM8.41817.0001CS 4558.3 CAD
EM8.41817.0001 EM8.41817.0010 CA8.41817.0001 CA8.41817.0010
Tris(2-methylphenyl)phosphine for synthesis, Sigma-Aldrich®
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 187…192 °C (0.05 mmHg)
Melting Pt: 125.5 °C
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now

Specification Test Results

Melting Point 124 - 127 °C
Molar Mass 304.38 g/mol
Boiling Point 238 °C (15 hPa)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR