3-Mercapto-1,2-propanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol

M-Clarity™ quality level = MQ200

8.41705.0250 8.41705.1000 8.41705.0250 8.41705.1000
EM8.41705.0250CS 6609.14 CAD
EM8.41705.0250 EM8.41705.1000 CA8.41705.0250 CA8.41705.1000
3-Mercapto-1,2-propanediol for synthesis, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
MW: 108.16 g/mol
Boiling Pt: 118 °C (1013 hPa)
Density: 1.247 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,09471

Order Now

Specification Test Results

Molar Mass 108.15 g/mol
Boiling Point 118 °C (6.7 hPa)
Flash Point 110 °C
Density 1.247 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR