2-tert-Butylhydroquinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41424.0100 8.41424.0500 8.41424.0100 8.41424.0500
EM8.41424.0100CS 5681.68 CAD
EM8.41424.0100 EM8.41424.0500 CA8.41424.0100 CA8.41424.0500
2-tert-Butylhydroquinone for synthesis, Sigma-Aldrich®
2-tert-Butylhydroquinone
Formula: C₁₀H₁₄O₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 128 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002344
CAS Number: 1948-33-0
EINECS: 217-752-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Melting Point 127 - 129 °C
Molar Mass 166.21 g/mol
Boiling Point 295 °C (1013 hPa)
Flash Point 171 °C
Density 1.05 g/cm3 (20 °C)
Ignition Point 457 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR