Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.41331.0005
8.41331.0025
8.41331.0005
8.41331.0025
EM8.41331.0005CS
9867.1
CAD
EM8.41331.0005
EM8.41331.0025
CA8.41331.0005
CA8.41331.0025
Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Pentafluorobenzoyl chloride
Formula:
C₇ClF₅O Boiling Pt: 158 °C (1013 hPa) Density: 1.6 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000657 CAS Number: 2251-50-5 EINECS: 218-842-4 UN: 3265 ADR: 8,II |
Specification Test Results
Assay (GC, area%) | ≥98.0% |
Identity (IR) | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...