3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS, Sodium 3,5-Dichloro-2-hydroxybenzenesulfonate

M-Clarity™ quality level = MQ200

8.41262.0005 8.41262.0025 8.41262.0005 8.41262.0025
EM8.41262.0005CS 125455.16 CAD
EM8.41262.0005 EM8.41262.0025 CA8.41262.0005 CA8.41262.0025
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt for synthesis, Sigma-Aldrich®
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Formula: C₆H₃Cl₂NaO₄S
MW: 265.05 g/mol
MDL Number: MFCD00009798
CAS Number: 54970-72-8
EINECS: 259-416-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 265.05 g/mol
pH 7 (53 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR